missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, 0.5M buffer soln., pH 7.4, Thermo Scientific™
Click to view available options
Quantity:
500 mL
1 L
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Concentration | 0.5 M |
pH | 7.4 |
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | Not miscible or difficult to mix in water. Soluble in ethylene glycol (79.1mg/ml),methanol (26mg/ml),anhydrous ethanol (14.6mg/ml),95% ethanol (22mg/ml),DMF (14mg/ml),acetone (2mg/ml),ethyl acetate (0.5mg/ml),olive oil (0.4mg/ml),chloroform (0.05mg/ml),and alkaline solution. |
Chemical Name or Material | TRIS buffer soln. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'