missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.
Solubility
Soluble in DMSO, DMF.
Notes
Store at -20°C. Protect from heat. Incompatible with heat and oxidizing agents.
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.
Solubility
Soluble in DMSO, DMF.
Notes
Store at -20°C. Protect from heat. Incompatible with heat and oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 132°C to 143°C |
Color | White |
Quantity | 1 g |
Solubility Information | Soluble in DMSO,DMF. |
Formula Weight | 530.63 |
Percent Purity | ≥97% |
Physical Form | Solid |
Chemical Name or Material | Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'