missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tris(1-methoxy-2-methyl-2-propoxy)bismuth is used as metal organic precursors to prepare bismuth titanium oxide thin films, which finds application in ferroelectric random access memory.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon

Specifications
Specifications
Melting Point | 70°C |
Color | Yellow |
Quantity | 25 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethylcyclohexane. |
Formula Weight | 518.4 |
Percent Purity | 99.99% |
Physical Form | Crystalline |
Assay | (metals basis) |
Chemical Name or Material | Tris (1-methoxy-2-methyl-2-propoxy)bismuth |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction