missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%, Thermo Scientific Chemicals

Product Code. 11374487
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15492-52-1
Molecular Formula C33H57O6Yb
Molecular Weight (g/mol) 722.867
MDL Number MFCD00000034
InChI Key KACMHCBXMZUMQV-LWTKGLMZSA-K
Synonym 2,2,6,6-tetramethylheptane-3,5-dione; ytterbium 3+, ytterbium 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate, tris 2,2,6,6-tetramethyl-3,5-heptanedionato ytterbium iii-yb reo yb tmhd 3
PubChem CID 44473428
IUPAC Name (Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;ytterbium(3+)
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Yb+3]

Specifications

Melting Point 167°C to 169°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 722.86
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'