missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade Product Code.: 15470127

Thermo Scientific Chemicals Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade

Product Code. 15470127
10 g
Click to view available options
Quantity:
2 g
5 g
10 g
This item is not returnable. View return policy

Product Code. 15470127

Brand: Thermo Scientific Chemicals J60316.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For reducing disulfide bonds in peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Molecular Formula C9H12O6P
Molecular Weight (g/mol) 247.16
MDL Number MFCD00145469
InChI Key PZBFGYYEXUXCOF-UHFFFAOYSA-K
Synonym tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochlorideShow MoreShow Less
PubChem CID 2734570
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

Specifications

Melting Point 177°C to 178°C
Packaging Plastic bottle
Quantity 10 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO or water
Formula Weight 286.65
Percent Purity 98%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14L044 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberJ60316.06,J60316.09,J60316.04
Lot NumberP10L021 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberJ60316.06,J60316.09,J60316.04
Lot NumberN11L013 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberJ60316.06,J60316.09,J60316.04
Lot NumberZ17K018 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ60316.06,J60316.09,J60316.04
Lot NumberZ17K022 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ60316.06,J60316.09,J60316.04
compliance-icons
Product Identifier
  • Tris(2-carboxyethyl)phosphine hydrochloride, Molecular Biology Grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tris(2-carboxyethyl)phosphonium chloride
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-28

    New Offer
    Shop Now

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.