missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 10583182
58.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
51805-45-9
C9H12O6P
247.16
MFCD00145469
PZBFGYYEXUXCOF-UHFFFAOYSA-K
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
2734570
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
51805-45-9
C9H12O6P
247.16
MFCD00145469
PZBFGYYEXUXCOF-UHFFFAOYSA-K
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
2734570
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 04, IV, 3441
Merck Index 15, 9222
Solubility Information Solubility in water: soluble.
Formula Weight 286.65
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Tris(2-carboxyethyl)phosphine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'