Learn More
Thermo Scientific Acros Tris(2-thienyl)phosphine 98% m.p. 29 to 30°C Acros Organics
Brand: Thermo Scientific Acros 351800050
Tris(2-thienyl)phosphine 98% m.p. 29 to 30°C Acros Organics
Code : 29
Additional Details : CAS Number : 24171-89-9 Weight : 0.01000kg