missing translation for 'onlineSavingsMsg'
Learn More

Tris(3,6-dioxaheptyl)amine, 95%, Thermo Scientific Chemicals

Product Code. 11323137
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2'-deoxyadenosine and related benzimidazole 2'-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2′-deoxyadenosine and related benzimidazole 2′-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Incompatible strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70384-51-9
Molecular Formula C15H33NO6
Molecular Weight (g/mol) 323.43
MDL Number MFCD00010748
InChI Key XGLVDUUYFKXKPL-UHFFFAOYSA-N
Synonym tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine
PubChem CID 112414
IUPAC Name 2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine
SMILES COCCOCCN(CCOCCOC)CCOCCOC

Specifications

Melting Point <-100°C
Density 1.011
Boiling Point 163°C to 171°C (1 mmHg)
Flash Point 162°C (323°F)
Refractive Index 1.4486
Quantity 25 g
Beilstein 2369296
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 323.43
Percent Purity 95%
Chemical Name or Material Tris(3,6-dioxaheptyl)amine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'