missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents

Brand:  Fisher BioReagents BP1335-4

80.64 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10775494

  • £126.00 / 4 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TAE buffer solution - simply dilute as needed
  • 10X solution contains 0.4M tris-acetate and 0.01M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tris-Acetate-EDTA
64-19-7, 7732-18-5, 60-00-4, 77-86-1
4 L
Filtered through a 0.2-micron filter.
Poly CUBE
TAE
Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%).
10X Solution
Liquid
DNase free
DNase-, RNase- and Protease-Free
Protease free
Electrophoresis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers