missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAE Buffer, Tris-Acetate-EDTA, 25X Solution, Electrophoresis, Fisher BioReagents
Brand: Fisher BioReagents BP1330-1
This item is not returnable.
View return policy
Description
Nalgene Poly Bottle; 1L
- Save time and simplify your buffer preparation step by using Fisher BioReagents 25X TAE buffer solution - simply dilute as needed
- 25X solution contains 1M tris-acetate and 0.025M EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 24 volumes of ultrapure water, such as BP2819
- Tested for the absence of DNase, RNase, and protease
- Filtered, autoclaved, and functionally tested for use in electrophoresis applications
- TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
- Intended for laboratory research use only
Specifications
Tris-Acetate-EDTA | |
60-00-4, 6850-28-8, 77-86-1, 7732-18-5 | |
1 L | |
Filtered through a 0.2-micron filter. | |
Protease free | |
Electrophoresis |
25X Solution | |
Liquid | |
DNase free | |
Nalgene Poly Bottle | |
TAE | |
Tris (11.31%), EDTA Disodium Salt (0.87%), Acetic Acid, Sodium Salt (7.66%), and Hydrogen Chloride (3.74%). |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction