missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS (Base), BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 100 g |
Formula Weight | 121.14 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | TRIS (Base) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'