missing translation for 'onlineSavingsMsg'
Learn More

Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™

Product Code. 10103203
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Unit Size:
10kg
1kg
25kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10103203

Brand: Fisher BioReagents BP152500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

DNase-, RNase- and Protease-Free. | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

DNase-, RNase- and Protease-Free.

Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance 0.05 AU max. at 280nm
Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Crystalline Powder or Crystals
Quantity 500 g
DNase DNase free
Formulation 2-Amino-2-(Hydroxymethyl)-1,3-Propanediol, Tris (hydroxymethyl) Aminomethane, Tris Base, THAM, Trimethylol Aminomethane, Trometamol
Assay Percent Range ≥99.8 %
Identification Pass Test
Linear Formula H2NC(CH2OH)3
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 9951
Formula Weight 121.14
Percent Purity ≥99.8%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Base
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.