missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Unit Size:
10kg
1kg
25kg
500g
5kg
Description
DNase-, RNase- and Protease-Free.
Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
Specifications
Specifications
| Absorbance | 0.05 AU max. at 280nm |
| Melting Point | 168.5°C |
| Boiling Point | 219°C |
| Color | White |
| pH | 10.4 |
| Physical Form | Powder or Solid |
| Quantity | 10 kg |
| DNase | DNase free |
| Assay Percent Range | ≥99.8 % |
| Identification | Pass Test |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction