missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Buffer, 0.3M Solution (Molecular Biology), Fisher BioReagents
Click to view available options
Poly Bottle; 500mL
Description
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 0.3M, provides a convenient stock solution for these applications.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 77-86-1, 60-00-4 |
Color | Undesignated |
Concentration | 0.29 to 0.31 M |
pH | 10.1 to 11.1 |
Physical Form | Liquid |
Quantity | 500 mL |
DNase | DNase free |
Identification | Pass Test |
Molecular Formula | C4H11NO3 |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'