missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS-CAPS Buffer, 10X, Thermo Scientific™
For blot transfer
£50.83 - £89.51
Specifications
Chemical Name or Material | Buffer |
---|---|
Concentration | 10X |
For Use With (Application) | Blot transfer |
Physical Form | Liquid |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Buffer | |
10X | |
Blot transfer | |
Liquid |
Safety and Handling
Recommended Storage : Ambient temperatures