missing translation for 'onlineSavingsMsg'
Learn More
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals Product Code.: 10170292

Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals

Product Code. 10170292
5 g, Glass bottle
Click to view available options
Quantity:
500 mg
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10170292

Brand: Thermo Scientific Acros 318770050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51364-51-3
Molecular Formula C51H42O3Pd2
Molecular Weight (g/mol) 915.73
MDL Number MFCD00013310
InChI Key CYPYTURSJDMMMP-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium
PubChem CID 9811564
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 152.0°C to 155.0°C
Color Black to Purple
Elemental Analysis 96.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, thf, benzene, toluene
Formula Weight 915.7
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium(0)
Show More Show Less
compliance-icons
Product Identifier
  • Tris(dibenzylideneacetone)dipalladium(0)
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.