missing translation for 'onlineSavingsMsg'
Learn More
Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals Product Code.: 11349328

Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals

Product Code. 11349328
5 g
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11349328

Brand: Thermo Scientific Alfa Aesar 043237.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of aluminum nitrides

Preparation of aluminum nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32093-39-3
Molecular Formula C12H36Al2N6
Molecular Weight (g/mol) 318.425
MDL Number MFCD00269811
InChI Key JGZUJELGSMSOID-UHFFFAOYSA-N
Synonym tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminumShow MoreShow Less
PubChem CID 16688802
IUPAC Name N-[bis(dimethylamino)alumanyl]-N-methylmethanamine
SMILES CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C

Specifications

Melting Point 87°C to 89°C
Color Brown to White
Density 0.865 g/mL
Boiling Point 90°C (0.05 mmHg)(sublimation)
Flash Point 21°C (70°F)
Linear Formula Al2[N(CH3)2]6
Quantity 5 g
UN Number UN3131
Sensitivity Moisture sensitive
Formula Weight 318.42
Physical Form Powder
Chemical Name or Material Tris (dimethylamino)aluminum dimer
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR03F071 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number043237.03,043237.14,043237.06
Lot NumberY25F081 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number043237.03,043237.14,043237.06
Lot NumberW18K028 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog Number043237.03,043237.14,043237.06
Lot NumberP29E052 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043237.03,043237.06
Lot NumberR03F071 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043237.03,043237.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.