missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0 Product Code.: 10586441

Thermo Scientific Chemicals Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0

Product Code. 10586441
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 10586441

Brand: Thermo Scientific Chemicals 327345000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent, pH buffer
TRUSTED_SUSTAINABILITY

Specifications

CAS 38641-82-6
Color Colorless
pH 7.9 to 8.1 (25°C)
Physical Form Liquid
Quantity 500 mL
Molecular Formula C14H27N3O11
MDL Number MFCD00236359
Packaging HDPE Bottle
Beilstein 04, 303
Merck Index 14, 9772
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane
InChI Key VLEIUWBSEKKKFX-UHFFFAOYSA-N
SMILES NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 413.38
PubChem CID 16218629
Grade Molecular Biology
Chemical Name or Material Tris EDTA buffer, pH 8.0
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463497 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number327345000
Lot NumberA0455069 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number327345000
Lot NumberA0446592 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number327345000
Lot NumberA0390679 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number327345000
Lot NumberA0400230 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number327345000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.