missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TE Buffer, Tris-EDTA, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Brand: Fisher BioReagents BP2477-1
This item is not returnable.
View return policy
Description
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TE buffer solution - simply dilute as needed
- 10X solution contains 100mM tris and 10mM EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TE solution with 9 volumes of ultrapure water, such as BP2819
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- TE buffer is routinely used for resuspending nucleic acid samples
- Intended for laboratory research use only
Specifications
Tris-EDTA (TE), 10X Solution | |
77-86-1, 6381-92-6, 7647-01-0, 7732-18-5 | |
7.4 to 8.1 | |
1 L | |
Filtered through a 5-micron filter and autoclaved. | |
Protease free | |
Molecular Biology |
pH 7.4 | |
Colorless | |
Liquid | |
DNase free | |
Nalgene Poly Bottle | |
TE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TE Buffer, Tris-EDTA, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents > Nalgene Poly Bottle; 1L
Spot an opportunity for improvement?Share a Content Correction