Learn More
Tris(ethylenediamine)cobalt(III) chloride trihydrate, Thermo Scientific Chemicals
£38.70 - £142.00
Chemical Identifiers
CAS | 14883-80-8 |
---|---|
Molecular Formula | C6H30Cl3CoN6O3 |
Molecular Weight (g/mol) | 399.628 |
MDL Number | MFCD00211318 |
InChI Key | WBMVTRUQSMZIKW-UHFFFAOYSA-K |
Synonym | tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride |
PubChem CID | 73995044 |
IUPAC Name | cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate |
SMILES | C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493427
|
Thermo Scientific Alfa Aesar
A17211.09 |
10 g |
£38.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403437
|
Thermo Scientific Alfa Aesar
A17211.18 |
50 g |
£142.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14883-80-8 | |
399.628 | |
WBMVTRUQSMZIKW-UHFFFAOYSA-K | |
73995044 | |
C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3] |
C6H30Cl3CoN6O3 | |
MFCD00211318 | |
tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride | |
cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate |
Specifications
14883-80-8 | |
MFCD00211318 | |
WBMVTRUQSMZIKW-UHFFFAOYSA-K | |
cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate | |
73995044 | |
Tris(ethylenediamine)cobalt(III) chloride trihydrate |
C6H30Cl3CoN6O3 | |
tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride | |
C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3] | |
399.628 | |
399.64 (345.59 Anhydrous) |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only