Learn More
Tris-Glycine, 1X Solution (Electrophoresis), Fisher BioReagents
Brand:  Fisher Bioreagents BP2439-20
Code : 32
Additional Details : Weight : 20.05000kg
Product Code. 11443024
  • £492.00 / 20 L
Estimated Shipment: 04-10-2024
to see stock.
Description
Description
Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.
Specifications
Specifications
Tris-Glycine
56-40-6,77-86-1,7732-18-5
20 L
Filtered through a 0.2-micron filter.
Poly CUBE
Electrophoresis
1X Solution
8.3 to 8.5 (1X soln.)
Not Detected
Pass Test
Not Detected
Product Suggestions
Product Suggestions
Videos
SDS Documents