Learn More
Tris-Glycine-SDS, 10X Solution (Electrophoresis), Fisher BioReagents™
Brand: Fisher BioReagents BP1341-1
64.64 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Tris-Glycine-SDS, 10X Solution (Electrophoresis)
Tris-Glycine-SDS is commonly used as a buffer in SDS-PAGE of proteins.
Specifications
Tris-Glycine-SDS | |
77-86-1, 7732-18-5, 56-40-6, 151-21-3 | |
Liquid | |
DNase free | |
DNase-, RNase- and Protease-Free | |
Nalgene Poly Bottle | |
TG-SDS | |
0.25M Tris Base, 1.92M Glycine, and 1.0% (w/v) SDS. |
10X Solution | |
Undesignated | |
1 L | |
Filtered through a 0.2-micron filter. | |
Pass Test | |
Protease free | |
Electrophoresis |
Safety and Handling
- TG-SDS 10X Solution
Your input is important to us. Please complete this form to provide feedback related to the content on this product.