missing translation for 'onlineSavingsMsg'
Learn More
Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals Product Code.: 15835398

Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15835398 Shop All Thermo Scientific Products
500 mL, Nalgene bottle
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
This item is not returnable. View return policy

Product Code. 15835398

Brand: Thermo Scientific J22638.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Concentration 1 M
pH 8
Physical Form Solution
Quantity 500 mL
Packaging Nalgene bottle
Formula Weight 157.6
Grade Molecular Biology
Chemical Name or Material Tris-HCl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number234382 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ22638.AP,J22638.AE,J22638.K2
Lot Number241530 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ22638.AP,J22638.AE,J22638.K2
Lot Number201796 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ22638.AP,J22638.AE,J22638.K2
Lot Number232719 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ22638.AP,J22638.AE,J22638.K2
Lot Number231124 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberJ22638.AP,J22638.AE,J22638.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.