missing translation for 'onlineSavingsMsg'
Learn More

Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.), Fisher BioReagents™

£56.60 - £68.10

Chemical Identifiers

CAS 77-86-1, 1185-53-1, 7732-18-5
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris-HCl
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10031793
View Documents Promotion Details
Fisher BioReagents
BP1757-100
100 mL Poly Bottle
£56.60
100mL
Please to purchase this item. Need a web account? Register with us today!
10573145
View Documents Promotion Details
Fisher BioReagents
FLBP1757-500
500 mL Poly Bottle
£68.10
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.)

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.5 provide convenient and highly purified solutions.
Specifications

Chemical Identifiers

77-86-1, 1185-53-1, 7732-18-5
121.136
Tris-HCl
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1, 1185-53-1, 7732-18-5
0.96 to 1.04 M
Liquid
Filtered through a 0.2-micron filter.
C4H11NO3
Tris-HCl
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Tris Hydrochloride, 1M Solution, pH 7.0, pH 7.5, and pH 8.0
Undesignated
10
DNase free
Pass Test
Protease free
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
DNase-, RNase- and Protease-Free
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications