missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Description
For electrophoresis, molecular biology, and cell culture applications
Tris Hydrochloride is a buffering medium for electrophoresis, molecular biology, and cell culture applications. It may be combined with Tris Base to simplify Tris buffer preparation.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Absorbance | 0.05 max. (1M solution) at 280nm |
Melting Point | 150°C |
Color | White |
pH | 3.5 to 5.0 |
Physical Form | Solid |
Quantity | 1 kg |
DNase | DNase free |
Formulation | Tris (hydroxymethyl) Aminomethane, Hydrochloride |
Assay Percent Range | ≥99 % |
Identification | Pass Test |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Tris hydrochloride
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'