missing translation for 'onlineSavingsMsg'
Learn More
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents Product Code.: 10316893

Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents

Product Code. 10316893
500 g, Poly Bottle
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 10316893

Brand: Fisher BioReagents BP153500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For electrophoresis, molecular biology, and cell culture applications

Tris Hydrochloride is a buffering medium for electrophoresis, molecular biology, and cell culture applications. It may be combined with Tris Base to simplify Tris buffer preparation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane Hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Absorbance 0.05 max. (1M solution) at 280nm
Melting Point 150°C
Color White
pH 3.5 to 5.0
Physical Form Solid
Quantity 500 g
DNase DNase free
Assay Percent Range ≥99 %
Identification Pass Test
Linear Formula H2NC(CH2OH)3·HCl
Packaging Poly Bottle
Protease Protease free
Formula Weight 157.6
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Tris hydrochloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.