missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99.8%, for analysis, biochemical grade
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Absorbance | (40% aq. soln.) (1 cm cell) (10% aq. soln.) (1 cm cell), (10% aq. soln.) (1 cm cell) (40% aq. soln.) (1 cm cell) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm, 0.2 max. at 290nm |
Melting Point | >168.5°C |
Color | Brown to White |
pH | 10.0 to 11.5 (5% aq. soln.) |
Boiling Point | 219.0°C to 220.0°C (10.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.8% |
Packaging | Plastic bottle |
Linear Formula | (HOCH2)3CNH2 |
Quantity | 1 kg |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'