missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Pro
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
Absorbance | (0.1M in water), (0.1 M in water) at 0nm, 0.03 max. at 260nm |
Melting Point | >168.5°C |
Color | White |
pH | 10.0 to 11.7 (5% soln. at 25°C) |
Boiling Point | 219.0°C to 220.0°C (10.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.85% |
Packaging | Plastic Bottle |
Linear Formula | (HOCH2)3CNH2 |
Quantity | 1 kg |
Show More |
Safety and Handling
Product Identifier
- Tris(hydroxymethyl)aminomethane
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'