missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry

Product Code. 10649623
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance (40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
Melting Point >168.5°C
Color White
pH 10.0 to 11.5 (5% aq. soln.)
Boiling Point 219.0°C to 220.0°C (10.0 mmHg)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic drum
Molecular Formula C4H11NO3
Linear Formula (HOCH2)3CNH2
Quantity 10 kg
Beilstein 04, 303
Merck Index 15, 9951
Solubility Information Solubility in water: 550g/L (25°C). Other solubilities: 79.1g/L ethylene glycol- 26g/L methanol (25°C), 22g/L 95% ethanol- 14g/L dmf (25°C), 2g/L acetone- 0.5g/L ethyl acetate (25°C), 0.05g/L chloroform- <0.05g/L ccl4 (25°C)
Molecular Weight (g/mol) 121.14
Formula Weight 121.14
Percent Purity 99+%
Grade Biochemistry
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Product Identifier
  • Tris(hydroxymethyl)aminomethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.