missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane acetate salt, 99%

£27.14 - £245.22

Chemical Identifiers

CAS 6850-28-8
Molecular Formula C4H11NO3·C2H4O2
Molecular Weight (g/mol) 181.19
InChI Key PIEPQKCYPFFYMG-UHFFFAOYSA-N
Synonym tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
PubChem CID 16218782
ChEBI CHEBI:66869
IUPAC Name acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES CC(=O)O.C(C(CO)(CO)N)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11995281
View Documents
Thermo Scientific Chemicals
434500250
25 g Plastic bottle
£27.14
25g
Please to purchase this item. Need a web account? Register with us today!
11905291
View Documents
Thermo Scientific Chemicals
434501000
100 g Plastic bottle
£73.67
100g
Please to purchase this item. Need a web account? Register with us today!
11915291
View Documents
Thermo Scientific Chemicals
434505000
500 g Plastic bottle
£245.22
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

6850-28-8
181.19
tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
CHEBI:66869
CC(=O)O.C(C(CO)(CO)N)O
C4H11NO3·C2H4O2
PIEPQKCYPFFYMG-UHFFFAOYSA-N
16218782
acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

6850-28-8
White
C4H11NO3·C2H4O2
(1g/ml in water) Clear colorless
CC(=O)O.C(C(CO)(CO)N)O
181.19
CHEBI:66869
99%
Tris(hydroxymethyl)aminomethane acetate salt
0.5 max. (c=50, water) at 290nm
99%
tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
PIEPQKCYPFFYMG-UHFFFAOYSA-N
acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol
16218782
181.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 229-939-6

Product Identifier
  • Tris(hydroxymethyl)aminomethane acetate salt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.