missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure

£51.76 - £940.68

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H11NO3·HCl
Molecular Weight (g/mol) 157.6
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10093020
View Documents Promotion Details
Thermo Scientific Chemicals
228031000
100 g Plastic bottle
£51.76
100g
Please to purchase this item. Need a web account? Register with us today!
10421271
View Documents
Thermo Scientific Chemicals
228032500
250 g Plastic bottle
£84.92
250g
Please to purchase this item. Need a web account? Register with us today!
10358360
View Documents
Thermo Scientific Chemicals
228030010
1 kg Plastic bottle
£240.20
1kg
Please to purchase this item. Need a web account? Register with us today!
10032593
View Documents
Thermo Scientific Chemicals
228030051
5 kg Plastic drum
£940.68
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

1185-53-1
157.6
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
C(C(CO)(CO)N)O.Cl
C4H11NO3·HCl
MFCD00012590
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride

Specifications

1185-53-1
White
Authentic
C4H11NO3·HCl
MFCD00012590
(1% in water) Clear colorless
C(C(CO)(CO)N)O.Cl
157.6
157.6
Extra Pure
Tris(hydroxymethyl)aminomethane hydrochloride
150.0°C to 153.0°C
3.5 to 5 (1% soln. at 25°C)
99+%
(HOCH2)3CNH2·HCl
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
99+%
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-684-5

Product Identifier
  • Tris(hydroxymethyl)aminomethane hydrochloride

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only