missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry, Fisher Chemical™
£91.70 - £707.00
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
MDL Number | 12590 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
Synonym | tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Description
Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry
Chemical Identifiers
1185-53-1 | |
157.594 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
93573 | |
C(C(CO)(CO)N)O.Cl |
C4H12ClNO3 | |
12590 | |
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
1185-53-1 | |
White | |
Plastic powder jar | |
12590 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
Solid |
150°C | |
3.5 to 5.0 | |
C4H12ClNO3 | |
tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
157.6g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tris(hydroxymethyl)aminomethane hydrochloride
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tris(Hydroxymethyl)Methylamine Hydrochloride, 'Tris-HCl', for Biochemistry, Fisher Chemical™