missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)phosphine, 95%, Thermo Scientific Chemicals

Product Code. 10465375
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2767-80-8
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.08
MDL Number MFCD00055382
InChI Key JMXMXKRNIYCNRV-UHFFFAOYSA-N
Synonym tris hydroxymethyl phosphine, methanol, phosphinidynetris, trimethylolphosphine, phosphinidynetrimethanol, tris methanol phosphine, phosphinidynetrismethanol, unii-y6tg7wf7oq, y6tg7wf7oq, methanol, 1,1',1-phosphinidynetris, methanol, phosphinidynetri-7ci,8ci
PubChem CID 76001
IUPAC Name bis(hydroxymethyl)phosphanylmethanol
SMILES C(O)P(CO)CO

Specifications

Melting Point 48°C to 56°C
Color White to Yellow
Boiling Point 111.0°C to 113.0°C (2.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 124.08
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Tris(hydroxymethyl)phosphine
Show More Show Less
compliance-icons
Product Identifier
  • Tris(hydroxymethyl)phosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'