Learn More
Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%, Thermo Scientific Chemicals
£114.00 - £528.00
Chemical Identifiers
CAS | 103457-72-3 |
---|---|
Molecular Formula | C18H54ErN3Si6 |
Molecular Weight (g/mol) | 648.42 |
MDL Number | MFCD00271002 |
InChI Key | ZVNGWTZZRANJAO-UHFFFAOYSA-N |
Synonym | bis trimethylsilyl azanide; erbium 3+, tris n,n-bis trimethylsilyl amide erbium iii, acmc-20al2x, erbium iii bis trimethylsilyl amide, tris bis trimethylsilyl amino erbium, erbium tris bis trimethylsilyl amide, erbium 3+ tris bis trimethylsilyl azanide, erbium tris trimethyl-n-trimethylsilyl silanaminide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1 |
PubChem CID | 6098739 |
IUPAC Name | bis(trimethylsilyl)azanide;erbium(3+) |
SMILES | C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448620
|
Thermo Scientific Alfa Aesar
044421.03 |
1 g |
£114.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438620
|
Thermo Scientific Alfa Aesar
044421.06 |
5 g |
£528.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
103457-72-3 | |
648.42 | |
ZVNGWTZZRANJAO-UHFFFAOYSA-N | |
6098739 | |
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3] |
C18H54ErN3Si6 | |
MFCD00271002 | |
bis trimethylsilyl azanide; erbium 3+, tris n,n-bis trimethylsilyl amide erbium iii, acmc-20al2x, erbium iii bis trimethylsilyl amide, tris bis trimethylsilyl amino erbium, erbium tris bis trimethylsilyl amide, erbium 3+ tris bis trimethylsilyl azanide, erbium tris trimethyl-n-trimethylsilyl silanaminide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1 | |
bis(trimethylsilyl)azanide;erbium(3+) |
Specifications
103457-72-3 | |
Pink | |
[[(CH3)3Si]2N]3Er | |
UN3396 | |
bis trimethylsilyl azanide; erbium 3+, tris n,n-bis trimethylsilyl amide erbium iii, acmc-20al2x, erbium iii bis trimethylsilyl amide, tris bis trimethylsilyl amino erbium, erbium tris bis trimethylsilyl amide, erbium 3+ tris bis trimethylsilyl azanide, erbium tris trimethyl-n-trimethylsilyl silanaminide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1 | |
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3] | |
648.42 | |
648.41 | |
Powder |
166°C | |
C18H54ErN3Si6 | |
MFCD00271002 | |
Moisture sensitive | |
ZVNGWTZZRANJAO-UHFFFAOYSA-N | |
bis(trimethylsilyl)azanide;erbium(3+) | |
6098739 | |
98% | |
Tris[N,N-bis (trimethylsilyl)amide]erbium (III) |
Safety and Handling
GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H261-H314
missing translation for 'dotInformation' : Hazard Class: 4.3; Packaging Group: I
missing translation for 'tsca' : No
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only