missing translation for 'onlineSavingsMsg'
Learn More

Tris, Ultra Pure, 1kg, MP Biomedicals

Product Code. 11491672 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11491672

Brand: MP Biomedicals 04819623

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

77-86-1 Purity: 99.9% Heavy metals (as Pb): <<5 ppm O.D. (260mm) of a 5% saturated solution: <<0.1 pKa at 20°C = 8.1 Biological buffer reagent

TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1
Quantity 1 kg
Molecular Formula C4H11NO3
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.136
PubChem CID 6503
ChEBI CHEBI:9754
Grade Ultrapure
Chemical Name or Material Tris
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.