missing translation for 'onlineSavingsMsg'
Learn More
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals Product Code.: 11434993

Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals

Product Code. 11434993
500 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11434993

Brand: Thermo Scientific Alfa Aesar A12274.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Stable and incompatible with bases, reducing agents, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrateShow MoreShow Less
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point 150°C (in -2H2O)
Density 1.76
Odor Odorless
Quantity 500 g
Beilstein 6104939
Merck Index 14,8602
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 294.10 (258.07 Anhydrous)
Percent Purity 99%
Chemical Name or Material Trisodium citrate dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX29K034 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog NumberA12274.0E,A12274.0C,A12274.36
Lot Number10253406 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA12274.0E,A12274.0C,A12274.36
Lot NumberP25K029 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog NumberA12274.0E,A12274.0C,A12274.36
Lot NumberZ08J015 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberA12274.0E,A12274.0C,A12274.36
Lot NumberX25J031 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberA12274.0E,A12274.0C,A12274.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.