missing translation for 'onlineSavingsMsg'
Learn More
Trisodium citrate dihydrate, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 11357908

Trisodium citrate dihydrate, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11357908
500 g
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11357908

Brand: Thermo Scientific Alfa Aesar 036439.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents Cleaning, Furnishing Care Products, Laundry Dishwashing Products and cleaning automobile radiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrateShow MoreShow Less
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point 150°C-2H2O
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0%
Beilstein 6104939
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 294.10 (258.07 Anhydrous)
Grade ACS Reagent
Chemical Name or Material Trisodium citrate dihydrate, For analysis ACS

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L022 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog Number036439.36,036439.A3
Lot NumberM03L011 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number036439.36,036439.A3
Lot NumberM03L012 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number036439.36,036439.A3
Lot NumberZ13K015 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number036439.36,036439.A3
Lot NumberX29K034 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog Number036439.36,036439.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trisodium citrate dihydrate, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.