missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Tritest: γ1 FITC/γ1 PE/CD45 PerCP (CE/IVD)
Three-color direct immunofluorescence reagent used as an isotype (negative) control for flow cytometric immunophenotyping of erythorcyte-lysed whole blood.
Brand: BD Biosciences™ 342415
Additional Details : Weight : 0.05300kg
Description
- Clone X40 X40 2D1 (CE/IVD)
- BD TriTEST™ Control, γ1 fluorescein isothiocyanate (FITC)/γ1 phycoerythrin (PE)/CD45 peridinin chlorophyll (PerCP), is a three-color direct immunofluorescence reagent used as an isotype (negative) control for flow cytometric immunophenotyping of erythrocyte-lysed whole blood with TriTEST reagents on the Becton Dickinson family of flow cytometers.
- The TriTEST Control is used to determine the unstained lymphocyte population on a fluorescence 1 (FL1) vs fluorescence 2 (FL2) display and to determine any nonantigen-specific antibody binding (nonspecific staining) present, particularly that caused by Fc receptors.
- BDIS TriTEST Control is for in vitro diagnostic use with BD in vitro diagnostic TriTEST reagents.
- Contents:
- Specificity: Clone X40, Format: FITC, Isotype: IgG<sub>1</sub> K
- Specificity: Clone: X40, Format: PE, Isotype: IgG<sub>1</sub> K
- Specificity: CD45, Clone: 2D1, Format: PerCP, Isotype: IgG<sub>1</sub> K
Specifications
Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. | |
50 Tests | |
Immunofluorescence Flow Cytometry Test Reagents | |
CE/IVD |