Learn More
BD Biosciences™ Tritest: CD4 FITC/CD8 PE/CD3 PerCP (CE/IVD)
Three-color direct immunofluorescence reagent for determining percentages and absolute counts of mature human T lymphocytes (CD3+), helper/inducer (CD3+, CD4+) T lymphocytes, and suppresor/cytotoxic (CD3+, CD8+) T-lymphocytes in erythrocyte-lysed whole blood.
Brand: BD Biosciences™ 342414
Additional Details : Weight : 0.05300kg
Description
- Clone SK3 (Leu3a) SK1 SK7 (Leu-4) (CE/IVD)
- BD TriTEST™ CD4 fluorescein isothiocyanate (FITC)/CD8 phycoerythrin (PE)/CD3 peridinin chlorophyll protein (PerCP) is a three-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the absolute counts of mature human T lymphocytes (CD3+), helper/ inducer (CD3+CD4+) T lymphocytes, and suppressor/cytotoxic (CD3+CD8+) T lymphocytes in erythrocyte-lysed whole blood.
- When used with TruCOUNT™ Tubes, absolute counts of these populations can be enumerated from a single tube.
- BD TriTEST reagent and TruCOUNT Tubes can be used with the FACS Loader.
- Contents:
- Specificity: CD4, Clone SK3 (Leu3a), Format: FITC, Isotype: IgG<sub>1</sub> K, Entrez Gene ID: 920
- Specificity: CD8, Clone: SK1, Format: PE, Isotype: IgG<sub>1</sub> K, Entrez Gene ID: 925
- Specificity: CD3, Clone: SK7 (Leu-4), Format: PerCP, Isotype: IgG<sub>1</sub> K, Entrez Gene ID: 916
Specifications
Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration. | |
50 Tests | |
Immunofluorescence Flow Cytometry Test Reagents | |
CE/IVD |