missing translation for 'onlineSavingsMsg'
Learn More

Trithiocyanuric acid, 95%, Thermo Scientific Chemicals

Product Code. 11489376
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-16-4
Molecular Formula C3H3N3S3
Molecular Weight (g/mol) 177.258
MDL Number MFCD00006052
InChI Key WZRRRFSJFQTGGB-UHFFFAOYSA-N
Synonym trithiocyanuric acid, thiocyanuric acid, 1,3,5-triazine-2,4,6-trithiol, trismercaptotriazine, trimercaptocyanuric acid, cyanuric acid, trithio, usaf th-3, 1,3,5-triazine-2,4,6 1h,3h,5h-trithione, s-triazine-2,4,6-trithiol, 2,4,6-triazinetrithiol
PubChem CID 1268121
IUPAC Name 1,3,5-triazinane-2,4,6-trithione
SMILES C1(=S)NC(=S)NC(=S)N1

Specifications

Melting Point >300°C
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 177.27
Percent Purity 95%
Chemical Name or Material Trithiocyanuric acid
compliance-icons
Product Identifier
  • Trithiocyanuric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Trithiocyanuric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.