Learn More
Tritolyl phosphate, mixture of isomers, Thermo Scientific Chemicals
Description
Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, sulfuric acid, caustics, ammonia, aliphatic amines, alkanolamines, isocyanates, alkylene oxides, epichlorohydrin and nitromethane.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.167 |
Boiling Point | 265°C (10 mmHg) |
Flash Point | 250°C (482°F) |
Odor | Odorless |
Linear Formula | (CH3C6H4O)3P=O |
Quantity | 2500 mL |
UN Number | UN2574 |
Merck Index | 14,9763 |
Solubility Information | Miscible with organic solvents. Immiscible with water. |
Formula Weight | 368.37 |
Show More |
RUO – Research Use Only