missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Triton∣r X-100, Electrophoresis Reagent
Click to view available options
Quantity:
100 mL
2500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Yellow |
Content And Storage | Ambient temperatures |
Density | 1.061 g/mL |
Quantity | 100 mL |
Formula Weight | ca. 625 |
MDL Number | MFCD00132505 |
Melting Point | ∼5°C |
Odor | Mild |
InChI Key | JYCQQPHGFMYQCF-UHFFFAOYSA-N |
SMILES | CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
Show More |
Safety and Handling
Product Identifier
- TritonX-100, Electrophoresis Reagent
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : TritonX-100
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction