missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, MP Biomedicals

Product Code. 11471632 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11471632

Brand: MP Biomedicals 04807426

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

  • The principal uses of Triton™ X-100 is in industrial and household detergent applications and in emulsifying agents
  • It is employed in almost every type of liquid, paste, and powdered cleaning compound, ranging from heavy-duty industrial products to gentle detergents for fine fabrics
  • It is also an important ingredient of primary emulsifier mixtures used in the manufacture of emulsion polymers, stabilizers in latex polymers, and emulsifiers for agricultural emulsion concentrates, and wettable powders

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless to Yellow
Physical Form Viscous Liquid
Quantity 500 mL
Linear Formula (C2H4O)x . C14H22O
Solubility Information Miscible in Water, Polar Organic Solvents (alcohols, glycols, ethers, ketones, etc.) & Aromatic Hydrocarbons; Soluble in Inorganic Salt Solutions & Aqueous Mineral Acids; Insoluble in Aliphatic Hydrocarbons.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Formula Weight 625 avg
Chemical Name or Material TRITON™ X-100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triton X-100
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.