missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton™ X-405

£15.89 - £84.20

Specifications

Chemical Name or Material Triton™ X-405
Color Yellow
Name Note 70% Solution in Water
CAS 9002-93-1, 7732-18-5
Physical Form Liquid
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10043623
View Documents
Thermo Scientific Chemicals
215692500
250 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10733601
View Documents
Thermo Scientific Chemicals
215690010
1 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10514664
View Documents
Thermo Scientific Chemicals
215690025
2.5 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triton™ X-405
70% Solution in Water
Liquid
-4.0°C
>109°C
1.4360 to 1.4380
C16H26O2
15, 6850
JYCQQPHGFMYQCF-UHFFFAOYSA-N
250.38
1968.47
Yellow
9002-93-1, 7732-18-5
120.0°C
1.0800g/mL
Glass bottle
1.08
MFCD00132505
Solubility in water: soluble.
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
5590
≥65%
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Triton™ X-405

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.