missing translation for 'onlineSavingsMsg'
Learn More

Trityl Tetrakis(pentafluorophenyl)borate, 97%

Product Code. 10491733 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10491733

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136040-19-2 | C43H15BF20 | 922.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136040-19-2
Molecular Formula C43H15BF20
Molecular Weight (g/mol) 922.37
MDL Number MFCD03426981
InChI Key TZOSNOQHGGONMD-UHFFFAOYSA-N
Synonym triphenylmethylium tetrakis perfluorophenyl borate, trityl tetrakis pentafluorophenyl borate, trityl tetrapentafluorophenyl borate, methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-, triphenylcarbenium tetrakis pentafluorophenyl borate, methylium, triphenyl-, tetrakis 2,3,4,5,6-pentafluorophenyl borate 1-1:1, tetrakis 2,3,4,5,6-pentafluorophenyl boranuide; triphenylmethylium, methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-1:1, tritylium tetrakis pentafluorophenyl borate, triphenylmethyliumtetrakis pentafluorophenyl borate
PubChem CID 9832824
SMILES C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
IUPAC Name diphenylmethylbenzene;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Formula Weight 922.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Trityl tetrakis(pentafluorophenyl)borate
compliance-icons
Product Identifier
  • Trityl tetrakis(pentafluorophenyl)borate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.