missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Key Industrial Equipment™ Trolley, stainless steel with two shelves, 100kg capacity
Brand: Key Industrial Equipment™ 1159O054
This item is not returnable.
View return policy
Description
General purpose stainless steel trolley suitable for catering and hosiptals. Corrosion resistant stainless steel shelves and handles. Wipe clean hygienic design. Easy handling from either end.- Two shelves
- Load capacity: 100kg
- Dimensions (H x L): 900mm x 610mm
- Non-marking castors, diameter 100mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction