missing translation for 'onlineSavingsMsg'
Learn More

Trolox(R), 97%, Thermo Scientific Chemicals

Product Code. 10782831
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53188-07-1
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.29
MDL Number MFCD00006846
InChI Key GLEVLJDDWXEYCO-UHFFFAOYNA-N
Synonym trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm
PubChem CID 40634
ChEBI CHEBI:82625
IUPAC Name 6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid
SMILES CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O

Specifications

Melting Point 187.0°C to 194.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 18, V,7, 335
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, ethanol
Formula Weight 250.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Trolox*, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Trolox
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trolox(R), 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.