missing translation for 'onlineSavingsMsg'
Learn More

Tromethamine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15588244
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Packaging Poly Bottle
Quantity 500 g
Formula Weight 121.14
Grade Multi-Compendial/USP
Chemical Name or Material Tromethamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'