Learn More
Tropine DL-tropate, 99%
Beskrivning
Tropine DL-tropate is used as a reduces brain levels of acetylcholine, Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors, used in opthalmology to dilate the pupil and in efforts of resuscitation in the treatment of bradycardia and cardiac arrest.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Quantity | 25 g |
| Description | White |
| For Use With (Application) | Reduces brain levels of acetylcholine |
| Formula Weight | 289.37g/mol |
| Sensitivity | Light Sensitive |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
| Beilstein | 91260 |
| InChI Key | RKUNBYITZUJHSG-UHFFFAOYNA-N |
| SMILES | CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1 |
| Visa mer |
RUO – Research Use Only
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.