missing translation for 'onlineSavingsMsg'
Learn More

Tropone, 97%, Thermo Scientific Chemicals

Product Code. 11360064
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-80-0
Molecular Formula C7H6O
Molecular Weight (g/mol) 106.124
MDL Number MFCD00014331
InChI Key QVWDCTQRORVHHT-UHFFFAOYSA-N
Synonym tropone, 2,4,6-cycloheptatrien-1-one, tropon, unii-co48x7sufh, co48x7sufh, cycloheptatrienone, 1-cyclohepta-2,4,6-trienone, 2,4,6-cycloheptatriene-1-one
PubChem CID 10881
IUPAC Name cyclohepta-2,4,6-trien-1-one
SMILES C1=CC=CC(=O)C=C1

Specifications

Density 1.094
Boiling Point 91°C to 92°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 5 g
Beilstein 1902335
Formula Weight 106.12
Percent Purity 97%
Chemical Name or Material Tropone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'